missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS Core Protein
A cDNA sequence encoding the SARS Core was constructed and used to recombinantly synthesize the protein.
£173.00 - £1100.00
Specifications
Name | SARS SARS Core Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | SARS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944058
|
enQuireBio™
QP10499-100ug |
100 μg |
£173.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15964058
|
enQuireBio™
QP10499-500ug |
500 μg |
£660.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15954058
|
enQuireBio™
QP10499-1mg |
1 mg |
£1100.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SARS SARS Core Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
Untagged | |
SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 60mM NaCl and 50% glycerol. |