missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep TGF-alpha Protein

A DNA sequence encoding the Ovis aries (Sheep) TGF-alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6777-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015780

  • £1930.00 / 500µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

TGF-alpha Protein
Research Use Only
TGFA
Recombinant Protein
Yeast
ENSTSALSDP PVAAAVVSHF NDCPDSHTQF CFHGTCRFLL QEEKPACVCH SGYVGARCEH ADLLAVVAAS QKKQ
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Extracellular Domain
Sheep
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers