Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 488, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand:  Chromotek SRBAF488-1-10
 View more versions of this product
Product Code. 17353493
  • £76.00 / 10µL
Explore more special offers
Description
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Specifications
Recombinant VHH
CTK0101 and CTK0102
10 μL
Immunofluorescence, Western Blot, Flow Cytometry
Alpaca
Human, Rabbit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 488, Clone: CTK0101 and CTK0102, Proteintech ChromoTek > 10 μL