missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0105 and CTK0106, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£76.00 - £277.00
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0105 and CTK0106 |
Applications | Immunofluorescence, Western Blot |
Host Species | Alpaca |
Target Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17353473
|
Chromotek
SMS2BAF488-1-10 |
10 μL |
£76.00
10µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17363473
|
Chromotek
SMS2BAF488-1-100 |
100 μL |
£277.00
100µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot | |
Mouse |
CTK0105 and CTK0106 | |
Alpaca |