missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0103 and CTK0104, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£77.35 - £280.91
Specifications
| Antigen | Recombinant VHH |
|---|---|
| Clone | CTK0103 and CTK0104 |
| Applications | Immunofluorescence, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17333473
|
Chromotek
sms1AF647-1-10 |
10 μL |
£77.35
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17343473
|
Chromotek
sms1AF647-1-100 |
100 μL |
£280.91
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-mouse IgG1, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.
Tekniske data
| Recombinant VHH | |
| Immunofluorescence, Western Blot, Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
| CTK0103 and CTK0104 | |
| Monoclonal | |
| Alpaca | |
| Secondary |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel