missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0105 and CTK0106, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£76.00 - £277.00
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0105 and CTK0106 |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17393473
|
Chromotek
SMS2BAF647-1-10 |
10 μL |
£76.00
10µL |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17303483
|
Chromotek
SMS2BAF647-1-100 |
100 μL |
£277.00
100µL |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot | |
Alexa Fluor 647 | |
Mouse | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
CTK0105 and CTK0106 | |
Monoclonal | |
Alpaca | |
Secondary |