missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0107, Proteintech ChromoTek
Alpaca Monoclonal Antibody
£76.00 - £276.00
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0107 |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17313483
|
Chromotek
sms3AF647-1-10 |
10 μL |
£76.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17323483
|
Chromotek
SMS3AF647-1-100 |
100 μL |
£276.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-mouse IgG3, Fab-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of a Nanobody that binds to mouse IgG3 with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot | |
Alexa Fluor 647 | |
Mouse | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
CTK0107 | |
Monoclonal | |
Alpaca | |
Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0107, Proteintech ChromoTek