Learn More
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0104, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand: Chromotek SMSG1CYS2-2-500
Additional Details : Weight : 0.00240kg
Descripción
500 μg Nano-Secondary™ for mouse IgG1: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0104].
Especificaciones
Recombinant VHH | |
CTK0104 | |
500 μg |
Immunofluorescence, Western Blot, Flow Cytometry | |
Alpaca | |
Mouse |