Learn More
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0105, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand:  Chromotek SMSG2BCYS2-1-500
Product Code. 17333493
  • £561.00 / 500µL
Explore more special offers
Description
Description
500 μg Nano-Secondary™ for mouse IgG2b: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0105].
Specifications
Specifications
Recombinant VHH
Monoclonal
Unconjugated
500 μg
Mouse
Immunofluorescence, Western Blot, Flow Cytometry
CTK0105
Alpaca
Secondary
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Recombinant VHH Alpaca, anti-Mouse, Unconjugated, Clone: CTK0105, Proteintech ChromoTek > 500 μg