missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral CCI Fc Chimera (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7055-CC-025/CF
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Viral CCI Fc Chimera (CHO-expressed) Protein is derived from CHO. The Recombinant Viral CCI Fc Chimera (CHO-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
CCI Fc Chimera (CHO-expressed) | |
Chinese Hamster Ovary cell line,CHO-derived viral CCI protein, (N-terminus) Viral CCI (Met1-Val258) Accession &Num; P19063 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
52.6 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |