Learn More
Gilac™ Rectangular HACCP Container with Lid
Rectangular HACCP Container with Lid is a food container for large storage volumes; lid can support the weight of heavy loads.
Brand:  Gilac™ G119818
 View more versions of this product
Product Code. 17928471
  • £64.55 / Each
Explore more special offers
Description
Description
  • HACCP rectangular food container.
  • For large storage volumes, lid can support the weight of heavy loads.
  • Multipurpose container.
  • Rewritable identification cartridge for inventory tracking.
  • No gaps nor overlaps.
  • Machine washable.
  • Recessed handles for better handling.
  • Stackable without lid.
  • Compatible with all GILAC 4 and 6 wheel dollies.
Specifications
Specifications
Storage Tote
Yellow
9.2 gal.
HDPE
Rectangular
35 L