missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Red Natural Rubber Filter Ring with Flange

Brand:  Deutsch Neumann™ 2350032

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10424313

  • £1.30 / Each
In Stock
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Red
1Each
56 mm
Natural Rubber
1 Each
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers