missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Laboratory Tubing

Provides flexibility, tear strength and resilience for general laboratory usage. Deutsch Neumann™ Laboratory Tubing is manufactured from red natural rubber.

Brand:  Deutsch Neumann™ 3010609

 View more versions of this product

Product Code. 10088439

  • £53.00 / 25m
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Durometer 50 Shore A
  • Good elastic memory
Specifications

Specifications

ISO 9001:2008
Rubber
25 m
For Laboratory Use
6 mm
9 mm
1.5 mm
Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers