Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Provides flexibility, tear strength and resilience for vacuum applications. Deutsch Neumann™ Vacuum Quality Laboratory Tubing is manufactured from red natural rubber.
Brand:  Deutsch Neumann™ 3021020
 View more versions of this product
Product Code. 10303661
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Durometer 50 Shore A
  • Good elastic memory
  • Excellent choice for vacuum applications
Specifications
Specifications
ISO 9001:2008
Natural Rubber
25 m
For Laboratory Use
10 mm
20 mm
5 mm
Red