missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Conical One-Hole Stoppers

Made from red rubber.

Brand:  Kartell™ 0382500

 View more versions of this product

Product Code. 10567051

  • £12.95 / Pack of 25
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Made from red rubber.

  • Made from red rubber
Specifications

Specifications

3mm
25/Pk.
Red
20 mm
Tapered Stopper
24mm
1
14 mm
Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers