missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Conical 2 Hole Stopper

Red rubber stoppers suitable for high temperatures and for use in combination with a flask, bottle, or tube

Brand:  Kartell™ 0384800

 View more versions of this product

Product Code. 10440332

  • £18.95 / Pack of 5
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of red rubber
  • Conical shape
  • Features two holes
  • For general purpose laboratory use
Specifications

Specifications

6mm
45mm
2
34 mm
Rubber
Conical
6mm
25/Pk.
Red
45 mm
Tapered Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers