Learn More
Redox Buffer, 220mV, Reagecon™

Used for testing, calibration and verification of redox/ORP sensors. Redox Buffer, 220mV, Reagecon™ is a ready to use solution of iron salts, including potassium ferrocyanide and tripotassium hexacyanoferrate, for use with reactions with a redox potential of 220mV.
Brand: Reagecon 90565
Description
Redox (reduction–oxidation) reactions cause an electron transfer process that can be measured using a Redox or ORP (oxidation-reduction potential) electrode. Redox is an absolute measurement expressed in millivolts. During its working life, a Redox electrode undergoes no change of zero point or slope. Therefore, redox electrodes do not require calibration. A control standard controls the functionality of the sensing and reference electrode, and also controls the analyst's technique, environmental conditions and the operation of the measurement meter.Specifications
13943-58-3, 13746-66-2 | |
100°C | |
Redox and ORP Sensors | |
250 mL | |
Characteristic | |
Fully miscible in water |
0°C | |
Test ORP electrodes | |
Liquid | |
Iron salts | |
23 hPa at 20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.