missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Redox standard solution, Jenway™
missing translation for 'orderingAttributeHoverText'
Solution Type:
Redox Standard (200 mV)
Redox Standard (300 mV)
Description
- Storage solution for pH electrodes
- 500mL bottle
Specifications
Specifications
Quantity | 500 mL |
Solution Type | Redox Standard (200 mV) |
Chemical Name or Material | pH Buffer |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'