missing translation for 'onlineSavingsMsg'
Learn More

Redox standard solution, Jenway™

Product Code. 11480609
Click to view available options
Solution Type:
Redox Standard (200 mV)
Redox Standard (300 mV)
This item is not returnable. View return policy
This item is not returnable. View return policy

Protection and care for pH electrodes

  • Storage solution for pH electrodes
  • 500mL bottle

Specifications

Quantity 500 mL
Solution Type Redox Standard (300 mV)
Chemical Name or Material pH Buffer
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'