missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Refill Kit for Trolleys Sorbents
Shop All asecos Products
missing translation for 'orderingAttributeHoverText'
Type:
Chemical Spill Control Absorbent Storage Cart Refill Kit
Oil Spill Control Absorbent Storage Cart Refill Kit
Universal All Purpose Absorbent Storage Cart Refill Kit
missing translation for 'unitSize'
Each
Description
- content: 6 cushions, 3 socks, 20 mats, 3 pair protective gloves, 5 plastic bags
Specifications
Specifications
| Material | Heavy-duty Cloth |
| Type | Oil Spill Control Absorbent Storage Cart Refill Kit |
| Weight (English) | 13.23 lb. |
| Weight (Metric) | 6 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction