Learn More
Hanna Instruments™ Refillable Digital pH Electrode
Brand:  Hanna Instruments™ HI11310
 View more versions of this product
Product Code. 15178239
  • £651.36 / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
With Sensor Check and Matching Pin
Ceramic Single, 15 to 20 μL/h
0 to 13
1 m
3.5 mm Jack
Glass
-5°C to +100°C
9.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Refillable Digital pH Electrode >