missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Refillable Racks for Eclipse™ Refill Tips
GREENER_CHOICE

Product Code. 16661963
Click to view available options
Holds:
Pipette Tips (10 μL)
Pipette Tips (10 to 1250 μL)
Pipette Tips (10 to 250 μL)
Pipette Tips (100 to 1250 μL)
Pipette Tips (1000 to 1250 μL)
Color:
Assorted
Purple
Teal
Unit Size:
Pack of 10
Pack of 12
Pack of 6

Product Code. 16661963

Brand: Labcon™ 12809600069

Only null left
Add to basket
Add to basket

Each rack is designed to hold one refill of 96 tips.

  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • For use with Eclipse™ pipette tip refills
  • Holds one refill platform of 96 tips
  • Autoclavable
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP
Product Type Refillable
Autoclavable Autoclavable
Color Purple
Holds Pipette Tips (10 to 250 μL)
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Liquid Handling
No. per Pack 10/Pk.
Quantity 10 Racks/Pk., 10 Pk./Cs.
Shipping Condition Room Temperature
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.