missing translation for 'onlineSavingsMsg'
Learn More

Labcon Refillable Racks for Labcon Tips
GREENER_CHOICE

Product Code. p-7193995
Click to view available options
Holds:
Tips (10 to 1250μL)
Tips (Labcon 10μL)
Unit Size:
Pack of 10
Pack of 12

Product Code. 16621963

Brand: Labcon 12109600069

Only null left
Add to basket
Add to basket

For use with Eclipse™ pipette tip refills

  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Product Type Empty Refill Rack with Lid
Autoclavable Autoclavable
Color Purple
Holds Tips (10 to 1250μL)
Material Polypropylene
Positions Holds one refill platform of 96 tips
Sterility Non-sterile
For Use With (Application) Liquid Handling
Packaging 10 racks / Pk./10 Pk. / case
Quantity 10 racks / Pk.
Shipping Condition RT
Size 10-1250 μL tips
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.