Learn More
3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses
Product Code. 11975482
Blue
Click to view available options
Frame Color:
Blue
Bronze
Provides excellent coverage with fewer gaps where dangerous flying debris can damage the eyes. 3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses are especially designed to fit smaller head and face shapes.
  • Available in 2 lens colors
  • Slim frame design for younger or female users
  • Integrated side shield for added protection
  • Soft, padded adjustable nose bridge provides a comfortable fit
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses; Safety Spectacles
Frame Color Blue
No. per Pack 1 Ea.
Lens Material Polycarbonate
Product Content Correction
Product Title
3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses >