Learn More
Medline Scientific™ Reflex Trinocular Microscope
Brand: Medline Scientific™ 6400.0000M
This item is not returnable.
View return policy
Specifications
220/240 V | |
Plan Achromatic | |
220/240 V | |
WF 10X/16 mm | |
Trinocular, 30° Inclined |
Iris and Field | |
Reversed quadruple nosepiece with click stop | |
50/60 Hz | |
Halogen |