Learn More
Abeba™ Reflexor™ 5700 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 5700/44
Additional Details : Weight : 1.20000kg
Description
- CE EN ISO 20347:2012 O1 SRB
- Smooth leather white
- Slip resistance PU outsole sole
- 2 replaceable Reflexor™ insoles (article no. 3566/ 3569)
- Sanitized treated
- Antistatic
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 O1 SRB | |
Microfiber | |
44 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
5700 |