Learn More
Bellingham and Stanley™ Refractometer 0 to 30% starch Bellingham and Stanley
The Eclipse refractometer is a low cost, high quality hand-held refractometer robustly constructed for every day use.
Brand:  Bellingham and Stanley™ 45-26
Code : 04
Additional Details : Weight : 0.01000kg
Product Code. 12313529
  • £251.60 / Each
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
The Eclipse refractometer is a low cost, high quality hand-held refractometer robustly constructed for every day use.
Specifications
Specifications
Handheld Digital Refractometer
0 to 30 Brix