Learn More
Medline Scientific™ CETI 12kg Quartz Refractometer
Semi-automated Abbe refractometer featuring fast, digital read-out for rapid throughput. Medline Scientific™ CETI 12kg Quartz Refractometer is particularly useful in manufacturing and production.
Brand:  Medline Scientific™ 8400.0000M
Product Code. 12948514
  • £5285.00 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Semi-automated
  • Digital read-out
  • Rapid and accurate readout of RI, %Brix value, %Brix temperature
  • RS232 PC/printer interface
  • Supplied with two spare bulbs, two spare fuses, calibration set, and instruction manual with correction and dispersion tables
Specifications
Specifications
0 to 95 Brix, 0 to 90 Brix ATC
RS232
12 kg
15°C to 50°C
220/240 V
1.3000 to 1.7000
Two spare bulbs, two spare fuses, calibration set, and instruction manual with correction and dispersion tables
370 x 470 x 240 mm
220/240 V 50/60 Hz with on/off switch (Euro connector)
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Datasheets
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Medline Scientific™ CETI 12kg Quartz Refractometer > Refractive index: 1.3000 to 1.7000nD, 0 to 95% Brix (Bx), and 0 to 90% Brix temp. Compensated (Bx-TC)