missing translation for 'onlineSavingsMsg'
Learn More

Refractory Metals, plasma standard solution, Specpure™, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL, Thermo Scientific Chemicals

£183.96 - £442.86

Specifications

Chemical Name or Material Refractory Metals plasma standard solution
Name Note Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL
MDL Number MFCD00151265
UN Number UN3264
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044270.AE
100 mL
£183.96
100mL
Please to purchase this item. Need a web account? Register with us today!
11450700
View Documents
Thermo Scientific Alfa Aesar
044270.AP
500 mL
£442.86
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Refractory Metals plasma standard solution
MFCD00151265
Liquid
Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL
UN3264
Specpure™
Videos
Safety and Handling

Safety and Handling

Harmful if inhaled, May be corrosive to metals, Causes severe skin burns and eye damage

Acute toxicity (category 4), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Refractory Metals, plasma standard solution, Specpure™, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr at 100μg/mL, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.