Learn More
Techne™ Refrigerated Bath
Handles wide range of temperatures
Brand:  Techne™ 12212-02
 View more versions of this product
Product Code. 11543769
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Built-in refrigeration for efficient cooling
  • Robust stainless steel construction makes for easy cleaning
Spezifikation
Spezifikation
12 L
35°C to 100°C
RB-12A
230 V
20°C at 240W, 0°C at 240W, -10° C at 230W
Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty.
3 Years
50/60 Hz
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote