Learn More
Grant Instruments™ Refrigerated Immersion Coolers
GREENER_CHOICE
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
Brand:  Grant Instruments™ C2GR
 View more versions of this product
Product Code. 11476487
  • £3090.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
  • C1GR 0ºC to 40ºC
  • The cooling coil can be continuously immersed in liquids up to 100ºC with the cooler switched off and may be used to cool liquid down from 100ºC but it is not designed for continuous operation above 40ºC.
  • C2GR: -15ºC to 40ºC
Specifications
Specifications
460 x 305 x 225 mm (D x W x H)
-15°C to +40°C
120/230 V
400w at 20°C, 320w at 0°C, 170w at -10°C
925 mm
120/230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products