missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 0.5 L
Plug-and-play tangential flow filtration cassette that is made from regenerated cellulose, for ultrafiltration and diafiltration of intermediate sample volumes.
Specifications
Filter Type | Ultrafiltration |
---|---|
Format | Cassette |
For Use With (Equipment) | Peristaltic Pump |
Material | Polycarbonate |
Material (Membrane) | Regenerated Cellulose |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
30178732
|
Sartorius
VF-S050H0030-IV |
30 kDa |
N/A
|
N/A | |||||
30178723
|
Sartorius
VF-S050H0005-IV |
5 kDa |
N/A
|
N/A | |||||
30178737
|
Sartorius
VF-S050H0002-IV |
2 kDa |
N/A
|
N/A | |||||
30178718
|
Sartorius
VF-S050H0300-IV |
300 kDa |
N/A
|
N/A | |||||
30178735
|
Sartorius
VF-S050H0010-IV |
10 kDa |
N/A
|
N/A | |||||
30178721
|
Sartorius
VF-S050H0100-IV |
100 kDa |
N/A
|
N/A | |||||
Description
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: Regenerated cellulose
- Recommended feed volume: 100 to 500 mL
Specifications
Ultrafiltration | |
Peristaltic Pump | |
Regenerated Cellulose | |
Non-sterile | |
0.5 L |
Cassette | |
Polycarbonate | |
0.1 to 0.5 L | |
TFF cassette |