missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 1 L
Plug-and-play tangential flow filtration cassette that is made from regenerated cellulose, for ultrafiltration and diafiltration of intermediate sample volumes.
£201.00
Specifications
Filter Type | Ultrafiltration |
---|---|
Format | Cassette |
For Use With (Equipment) | Peristaltic Pump |
Material | Polycarbonate |
Material (Membrane) | Regenerated Cellulose |
Product Code | Brand | MWCO | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | ||||||
30178711
|
Sartorius
VF-S050H0002-SV |
300 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
30178731
|
Sartorius
VF-S050H0005-SV |
100 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
30178715
|
Sartorius
VF-S050H0010-SV |
30 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
30178739
|
Sartorius
VF-S050H0030-SV |
10 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
30178724
|
Sartorius
VF-S050H0100-SV |
5 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
30178740
|
Sartorius
VF-S050H0300-SV |
2 kDa |
£201.00
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
Description
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: Regenerated cellulose
- Recommended feed volume: 500 to 1000 mL
Specifications
Ultrafiltration | |
Peristaltic Pump | |
Regenerated Cellulose | |
Non-sterile | |
1 L |
Cassette | |
Polycarbonate | |
0.5 to 1 L | |
TFF cassette |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 1 L