Learn More
Regular Unleaded Gasoline, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep S-RG-20K
Code : NEW
Additional Details : CAS Number : 86290-81-5 Weight : 0.05000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Regular Unleaded Gas-87octane | 86290-81-5 | 20,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Regular Unleaded Gasoline (High Level) | |
2mL amber prescored glass ampule | |
98.0 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Methanol-P&T |
86290-81-5,67-56-1 | |
1 mL | |
98.0 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Safety and Handling
- Regular Unleaded Gasoline (High Level)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : gasoline, methanol
Product intended for laboratory use only