missing translation for 'onlineSavingsMsg'
Learn More

Reichert™ Rhino Clinical Handheld VET360 Refractometer

Rugged portable refractometer for urine specific gravity and serum/plasma protein readings

Brand:  Reichert™ 137536l0

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 10655725

  • £248.71 / Each
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Includes: Manual has full set of conversion tables for protein, urine solids, and refractive index.

Description

Description

Rugged portable refractometer for urine specific gravity and serum/plasma protein readings

  • IP67 rated water resistant, dustproof, and impact resistant
  • Determines urine specific gravity and protein concentration in plasma, serum, and peritoneal fluid
  • Automatic temperature compensation provides accurate readings regardless of ambient temperature
  • Corrects all readings back to standard reference temperature of 20°C (68°F)—provides much greater accuracy than urinometers or reagent strips
  • Includes special scales for large and small animals
  • Completely sealed for indoor or outdoor use
  • Provides readings from 16° to 32°C (61° to 90°F)
To Use:
  • Place drop of sample on prism
  • Hold unit perpendicular to light source
  • Look into focusable eyepiece

Specifications

Specifications

Handheld Clinical VET360 Refractometer
±0.001 sp. gr., ±0.2g/100mL Serum Protein
Place drop of sample on prism, hold unit perpendicular to light source, look into focusable eyepiece
IP67
1.6 in.
9.2 in.
1.6 in.
1 to 1.06 Urine Specific Gravity, 2 to 14 g/100 mL Serum Protein
0.001 sp. gr., 0.1g/100mL Serum Protein
Manual has full set of conversion tables for Protein, Urine solids and refractive index
9.2 x 1.6 x 1.6 in. (233 x 40.6 x 40.6 mm)
40.6 mm
233 mm
40.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers