missing translation for 'onlineSavingsMsg'
Learn More

Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers

Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant

£258.00 - £264.00

Specifications

Type Handheld Ultra-durable Refractometer
For Use With (Application) Food and beverage, chemical, pharmaceutical and industrial cutting fluids
View More Specs

Products 3
Product Code Brand Accuracy Scale Range Increments Refractive Index Price Quantity & Availability  
Product Code Brand Accuracy Scale Range Increments Refractive Index Price Quantity & Availability  
10309714
View Documents
Reichert™
137531L0
±0.25 0 to 50 Brix 0.25% 1.333 to 1.42
£258.00
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
10546475
View Documents
Reichert™
137530L0
±0.2 0 to 30 Brix 0.2 and 0.5 1.333 to 1.3811
£258.00
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
10481825
View Documents
Reichert™
137533L0
±0.2 50 to 90 Brix 0.2% 1.4201 to 1.5177
£264.00
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
Description

Description

Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant

  • Automatic Temperature Compensation eliminates the need for temperature correction charts
  • Optics and instrument scales are permanently sealed inside the refractometer body
  • Magnified scales have easy-to-read markings and divisions for true readings and accurate results
  • Models are offered with Brix scales of 0-30, 0-50 and 50-90%

Order Info

Rhino Model for Industrial Fluid Testing is also available; see Cat. No. RE-10440.

Specifications

Specifications

Handheld Ultra-durable Refractometer
Food and beverage, chemical, pharmaceutical and industrial cutting fluids
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.