Learn More
Reid Vapour Pressure Standard ASTM D 4953A, 323A (Nominal Value: 84.41 kPa), Paragon Scientific™
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Vapour Pressure (DVPE), ASTM D5191
Brand: Paragon Scientific CRM-VPGA
Additional Details : Weight : 0.70000kg Transport : UN number : 1203 Chem class : 3 Pack group : II
Description
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified by Round Robin Method by independant test laboratories
- Ensures good interlaboratory correlation
- Supplied in tamper evident UN compliant security pack
- Pack size 250 mL
- 12 months shelf life
Specifications
UN 1203 |
Safety and Handling
- 250ML CRM-VPGA Fuels Testing Certified Reference Material
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use media other than water to extinguish.
- P391-Collect spillage.
- P273-Avoid release to the environment.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : gasoline
Nominal values quoted. Certified Values will vary from batch to batch.