missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reinecke Salt, ACS Reagent, ≥93.0%, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 272°C |
Physical Form | Solid |
Quantity | 100 g |
Linear Formula | NH4[Cr(NH3)2(SCN)4] |
UN Number | NONH for all modes of transport |
Formula Weight | 336.43g/mol |
Percent Purity | ≥93.0% |
Grade | ACS Reagent |
Packaging | Glass Bottle |
Chemical Name or Material | Reinecke Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'