missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, Honeywell™

Product Code. 15618070
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for use as a reagent for choline determination

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
MDL Number MFCD00066638

Specifications

Melting Point 268°C to 272°C
Physical Form Solid
Quantity 100 g
Linear Formula NH4[Cr(NH3)2(SCN)4]
UN Number NONH for all modes of transport
Formula Weight 336.43g/mol
Packaging Poly bottle
Chemical Name or Material Reinecke salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'