Learn More
BINDER™ Reinforced Rack
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 720, KBF/KBF PRO 720
KB/KBF/KB PRO/KBF PRO 260/470
KBF 130, KBF PRO 130
Unit Size:
Each
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
Specifications
| For Use With (Equipment) | KB/KB PRO 1060, KBF/KBF PRO 1060 |
| Material | Stainless Steel |
| Type | Rack |