Learn More
Memmert™ Reinforced Stainless Steel Shelf
Product Code. p-7220951 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP750eco
IPP1060eco
IPP750eco
Unit Size:
Each
Product Code. 17130255
Brand: Memmert™ IPP750ecoB32191
Only null left
Add to basket
Add to basket
Additional reinforced stainless steel shelf for reinforced chamber option
  • With guide bars and fixing screws
  • Requires option K1
  • Consider max. loading of chamber; max. loading 60 kg
Specifications
For Use With (Equipment) IPP750eco
Material Stainless Steel