missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relative Humidity Calibration Standards, Rotronic™
£54.03 - £80.50
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
IUPAC Name | lithium(1+) chloride |
SMILES | [Li+].[Cl-] |
Product Code | Brand | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Price | Quantity & Availability | |||||
13174947
|
Rotronic
EA80SCS |
Lithium chloride Concentration 10 to 20% | |||||||
13114957
|
Rotronic
|
Lithium chloride Concentration 25 to 50% |
£80.50
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
[Li+].[Cl-] |
ClLi | |
MFCD00011078 | |
lithium(1+) chloride |
Specifications
7447-41-8 | |
ClLi | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium(1+) chloride | |
Odorless | |
Liquid |
Humidity Meters | |
MFCD00011078 | |
[Li+].[Cl-] | |
42.39 | |
130°C |