missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Pipet Tips Reload Pack for Pipetman™

Reload your standard Pipetman™ diamond tip pipet tip box wiimply and quickly with a full pack of new tips. Gilson™ Pipet Tips Reload Pack for Pipetman™ are autoclavable.

£34.30 - £56.95

Specifications

Description Pipet tip
Volume (Metric) 100 to 1000 μL
Autoclavable Autoclavable
For Use With (Application) Liquid handling
Format Refill
View More Specs

Products 2
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
15297783
View Documents Promotion Details
Gilson™
F167014
D1000
£34.30
Pack of 576
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15277783
View Documents Promotion Details
Gilson™
F167012
DL10
£56.95
Pack of 960
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

  • Quickest way to reload empty box (sold separately)
  • Uses 70% less plastic
  • Recyclable
  • Easy to store
  • Affordable and convenient
  • Fully autoclavable
  • Available in three sizes:
    • 0.1 - 2.0μL
    • 2 - 2000μL
    • 100 - 10000μL
Specifications

Specifications

Pipet tip
Autoclavable
Refill
Polypropylene
100 to 1000 μL
Liquid handling
Yes
Videos
SDS
Documents

Documents

Product Certifications