missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25371-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis.Specifications
REM2 | |
Polyclonal | |
Unconjugated | |
REM2 | |
GTP binding protein REM 2, REM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161253 | |
-20°C | |
Liquid |
Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IYK8 | |
REM2 | |
REM2 Fusion Protein Ag21991 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |