Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Brand: Thermo Scientific™ R30955101
118.80 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
Specifications
E. coli Polyvalent 4 (types O44, O112, O124, O142) | |
Agglutinating Sera | |
2 mL |
E. coli | |
2 mL/Vial |
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent > E. coli Polyvalent 4 (types O44, O112, O124, O142)
Spot an opportunity for improvement?Share a Content Correction