missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Brand: Thermo Scientific™ R30858101
132.18 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Positive | |
Salmonella O Antigen | |
Polyvalent | |
2 mL/Vial | |
Salmonella O |
Salmonella O Polyvalent (group A - G) | |
Agglutinating Sera | |
Group A-G | |
2 mL |