Learn More
Thermo Scientific™ Agglutinating Sera, Salmonella O
Description
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
| Control Sets | Positive |
| Description | Factor 15 (group E2) |
| Detectable Analytes | Salmonella O Antigen |
| Type | Agglutinating Sera |
| Organism Group | Group E2 |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
| Target Organism Class | Salmonella O |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.