Learn More
Thermo Scientific™ Vibrio cholerae Polyvalent Agglutinating Sera
Use Vibrio cholerae Polyvalent Agglutinating Serum for serological identification of V. cholerae O1 for epidemiological and diagnostic purposes.
Brand: Thermo Scientific™ R30165001
205.16 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Serologically identify Vibrio cholerae O1 by slide agglutination technique for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Vibrio cholerae Polyvalent Agglutinating Serum. Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens.
Vibrio cholerae serogroup O1 is associated with most epidemic outbreaks of cholera. The O-serogrouping is based on antibodies that recognize structural differences in the heat stable O-antigen (lipopolysaccharide) of the bacteria. V. cholerae O1 and V. cholerae non-O1 isolates can be distinguished on the basis of simple agglutination tests.
- Ready to use - Prepared reagent
- Easy to store - Store at 2-8° C
- Easy to interpret -
- Test suspension - Agglutination should be strong and clearly visible within one minute.
- Control suspension - No visible agglutination.
Vibrio cholerae Polyvalent Agglutinating Sera provides 2mL of rabbit serum containing antibodies to Vibrio cholerae, preserved with 0.5% phenol and supplied ready to use in a dropper bottle. Each dropper bottle contains sufficient sera for 40 to 50 tests.
This polyvalent serum is intended for use in slide agglutination screening tests.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
2°C to 8°C | |
Yes | |
Vial | |
2 mL |
Vibrio cholerae Polyvalent | |
V. cholerae | |
2 mL/Vial |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.