Learn More
Thermo Scientific™ Remel™ BactiCard™ E. coli
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
Marca: Thermo Scientific™ R21108
189.86 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Detalles adicionales : CAS Number : 109-86-4 Weight : 0.10000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Descripción
Thermo Scientific™ Remel BactiCard E. coli is used for presumptive identification of Escherichia coli using enzyme technology.
CPT Code: 87147
CLIA Complexity: Non-waived
- Two separate test circles for MUG and Indole
- Results after 15 minutes of incubation
- Positive MUG test displays fluorescence under longwave ultraviolet light
- Positive Indole test evidenced by a color change to redv
- If indole and MUG are positive, then E. coli is present
Includes:
Test cards, rehydrating fluid, MUG reagent, and applicator sticks.
Especificaciones
2°C to 8°C | |
E. coli | |
1 Kit | |
15 min. |
BactiCard™ E. coli | |
BactiCard | |
25 Tests/Kit |
Seguridad y manipulación
- Danger
Clasificación
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Indicaciones de peligro
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Consejos de prudencia
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
For in vitro diagnostic use.