missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BactiStaph™ Latex Agglutination Test Kit
Detect clumping factor and protein A associated with the identification of Staphylococcus aureus while distinguishing S. aureus from other staphylococci.
£367.00 - £535.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | Positive,Negative |
Description | BactiStaph Latex Agglutination Test Kit |
CE Marker | Yes |
Detectable Analytes | Clumping Factor and Protein A |
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
12609937
|
Thermo Scientific™
R21143 |
BactiStaph Latex Agglutination Test Kit | 450 Tests/Kit | 2°C to 8°C |
£535.00
450 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12619937
|
Thermo Scientific™
R21144 |
BactiStaph Latex Agglutination Test Kit | 150 Tests/Kit | 2°C to 8°C |
£367.00
150 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific™ BactiStaph™ Latex Agglutination Test Kit.
Use this rapid test utilizing latex particles which are capable of detecting both clumping factor and protein A simultaneously. Agglutination of white latex particles is visible to the unaided eye within 60 seconds and represents a positive reaction.Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
BactiStaph Latex Agglutination Test Kit | |
Clumping Factor and Protein A | |
Latex reagent, positive control, negative control, disposable slides, stirring sticks | |
Staphylococcus aureus |
Positive,Negative | |
Yes | |
Staphylococcus aureus Test Kit | |
Latex |